Anti-HSP60 Mouse Monoclonal Antibody
Leverandør: ENZO LIFE SCIENCES
ADISPA829D ADISPA829F
ENZOADISPA829DEA 3425 NOK
ENZOADISPA829D ENZOADISPA829F
Anti-HSP60 Mouse Monoclonal Antibody
Antistoffer
Hsp60 is a member of the chaperonin family of heat shock proteins, with homologs functioning in the cytosol and mitochondria to fold nascent and aggregated proteins. Hsp60 is the eukaryotic homolog of the E. coli GroEL protein, and forms a multimeric complex in the mitochondria with Hsp10 (Cpn10) to form a large central cavity in which ATP-dependent protein folding takes place. TRiC/CCT, a eukaryotic relative of Hsp60, is expressed in the cytosol and participates in the folding of actin and tubulin substrates, but lacks any association with an Hsp10-like co-factor.