Anti-HMOX1 Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ADISPA895D ADISPA895F
ENZOADISPA895DEA 2950 NOK
ENZOADISPA895D ENZOADISPA895F
Anti-HMOX1 Rabbit Polyclonal Antibody
Antistoffer
Heme Oxygenase-1 (HO-1) also known as Hsp32, is the inducible isoform of heme oxygenase that catalyzes the NADPH, oxygen, and cytochrome P450 reductase dependent oxidation of heme to carbon monoxide, ferrous iron and biliverdin which is rapidly reduced to bilirubin.

  • Dilution range: Western blot (1:1,000, ECL)
  • Detects a band of ~32kDa by western blot

Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rabbit, Rat, Sheep
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol HMOX1 HMOX1
Antigen-navn Heme Oxidase 1 Heme Oxidase 1
Environmentally Preferable
Antigen synonymer Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1 Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
Antistofftype Primary Primary
Klonalitet Polyclonal Polyclonal
Konjugering Unconjugated Unconjugated
Reaktivitet Rabbit,Sheep,Human,Dog,Rat,Mouse
Vert Rabbit Rabbit
Isotype
Western blot Yes Yes
ImmunoKjemi Yes Yes
Flowcytometri Yes Yes
Immunogen Recombinant rat HO-1 (Hsp32) lacking the membrane spanning region Recombinant rat HO-1 (Hsp32) lacking the membrane spanning region
Skjema Liquid Liquid
Rensing Purified by Protein A affinity chromatography Purified by Protein A affinity chromatography
Lagringsbuffer PBS containing 50% glycerol and 0,09% sodium azide PBS containing 50% glycerol and 0,09% sodium azide
Lagringstemp –20 °C –20 °C
Frakttemp. Shipped on blue ice Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR