Human recombinant Hsp47 H Rec

Leverandør: ENZO LIFE SCIENCES

ADISPP535D ADISPP535F
ENZOADISPP535DEA 3200 NOK
ENZOADISPP535D ENZOADISPP535F
Human recombinant Hsp47 H Rec
Proteiner

Produced in E. coli.

Bestill nå

SPESIFIKASJONER

Protein/peptide name Hsp47 H Rec
Environmentally Preferable
Protein/peptide type Recombinant
Arter Human
Kilde
Renhet >90% (SDS-PAGE)
Formulering Liquid, in 30 mM TRIS, pH 8,0, containing 100 mM sodium chloride, 1 mM DTT, and 0,4 M arginine
Molekylvekt ~47 kDa
Rensing Purified by multi-step chromatography

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR