Anti-HSPD1 Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ADISPS875D ADISPS875F
ENZOADISPS875DEA 2650 NOK
ENZOADISPS875D ENZOADISPS875F
Anti-HSPD1 Rabbit Polyclonal Antibody
Antistoffer
GroEL and GroES are E. coli chaperonins, which are homologs of mitochondrial chaperonins Hsp60 and Hsp10. GroEL is a double toriodal assembly of 14 identical subunits which form two heptameric rings stacked back-to-back, with a cavity at each end. GroEL and its co-chaperonin GroES facilitate protein folding with an ATP-dependent mechanism.

  • Dilution range: Western blot (1:1,000, colorimetric)
  • Detects a band of ~60 kDa by western blot

Type: Primary
Antigen: GroEL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: E.coli
Bestill nå

SPESIFIKASJONER

Antigensymbol HSPD1
Environmentally Preferable
Antigen-navn Heat shock protein family D (Hsp60) member 1
Antigen synonymer GROEL|HSP65|HLD4|HSP-60|HuCHA60|HSP60|CPN60|SPG13
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Bacteria
Vert Rabbit
Isotype
Western blot Yes
Immunogen E. coli GroEL.
Skjema Liquid
Rensing purified by Protein A affinity chromatography
Lagringsbuffer PBS containing 50% glycerol and 0,09% sodium azide
Lagringstemp –20 °C
Frakttemp. Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR