Mouse;Rat obestatin

Leverandør: ENZO LIFE SCIENCES

ALX164005M001
ENZOALX164005M001EA 2800 NOK
ENZOALX164005M001
Mouse;Rat obestatin
Proteiner

Food intake modulator


Obestatin derives like ghrelin from proghrelin, but exhibits opposite physiological effects. Obestatin decreases food intake and body weight in rodents. Moreover, obestatin also counteracts ghrelin’s effect by decelerating gastric emptying and decreasing intestinal contractility. Obestatin binds to orphan G protein-coupled receptor GPR39. Amidation of the C-terminus seems to be important for obestatin’s activity.

Bestill nå

SPESIFIKASJONER

Protein/peptide name Obestatin
Environmentally Preferable
Arter Rat,Mouse
Kilde
Renhet >97% (HPLC)
Formulering Lyophilised

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR