Human MMP8 proenzyme (from neutrophils)
Leverandør: ENZO LIFE SCIENCES
ALX200421C005
ENZOALX200421C005EA 1725 NOK
ENZOALX200421C005
Human MMP8 proenzyme (from neutrophils)
Proteiner
MMP-8 is a glycoprotein containing complex N-linked oligosaccharides.
  • Hydrolyzes type I over type II, and III collagens
  • Activated MMP-8 is inhibited by TIMP-1 (Prod. No. ALX-200-426)
Isolated from stimulated human neutrophil granulocytes (buffy coat). Requires activation.
Bestill nå
SPESIFIKASJONER
Protein/peptide name MMP8 proenzyme
Environmentally Preferable
Arter Human
Kilde Neutrophils
Renhet >90% (SDS-PAGE)
Konsentrasjon 100 µg/ml
Formulering Liquid. In 50 mM TRIS-HCl, pH 7,0, containing 200 mM NaCl, 5 mM CaCl₂, 1 µM ZnCl₂, 0,05% BRIJ 35, and 0,05% sodium azide.