Human recombinant MMP2 proenzyme (from insect cells (Sf9))

Leverandør: ENZO LIFE SCIENCES

ALX201095C010
ENZOALX201095C010EA 8050 NOK
ENZOALX201095C010
Human recombinant MMP2 proenzyme (from insect cells (Sf9))
Proteiner

Produced in Sf9 cells.


Active MMP-2 is used to study the degradations of proteins of the extracellular matrix, including fibrillar collagens. MMP-2 is inhibited by tissue inhibitors of matrix metalloproteinases (TIMP), alpha2-macroglobulin and by chelators of divalent cations as EDTA or o-phenanthroline.

Bestill nå

SPESIFIKASJONER

Protein/peptide name MMP2 proenzyme
Environmentally Preferable
Protein/peptide type Recombinant
Arter Human
Kilde Insect cells (Sf9)
Renhet >95% (SDS-PAGE)
Konsentrasjon 0,2 mg/ml
Formulering Liquid. In 50 mM TRIS-HCl, pH 7,0, 200 mM NaCl, 5 mM CaCl₂, 0,05% Brij-35.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR