Recombinant DsbC (from E. coli)

Leverandør: ENZO LIFE SCIENCES

ALX201268C100
ENZOALX201268C100EA 4775 NOK
ENZOALX201268C100
Recombinant DsbC (from E. coli)
Proteiner

Produced in E. coli.

Bestill nå

SPESIFIKASJONER

Protein/peptide name DsbC
Environmentally Preferable
Protein/peptide type Recombinant
Arter
Kilde E. coli
Renhet >95% (SDS-PAGE)
Konsentrasjon 1 mg/ml
Formulering Liquid. In 20 mM TRIS, pH 7,5, containing 2 mM EDTA.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR