Human recombinant DESC1
Leverandør: ENZO LIFE SCIENCES
ALX2017511
ENZOALX2017511EA 6050 NOK
ENZOALX2017511
Human recombinant DESC1
Proteiner
Produced in Drosophila Schneider 2 (S2) cells. Recombinant human DESC1 (aa 44-423) is fused at the C-terminus to a V5-His-tag.