Human recombinant IL-3

Leverandør: ENZO LIFE SCIENCES

ALX2018200010
ENZOALX2018200010EA 3850 NOK
ENZOALX2018200010
Human recombinant IL-3
Proteiner

Produced in E. coli. Non-glycosylated protein, containing 133 amino acids.


  • Carrier-free

New Product

Bestill nå

SPESIFIKASJONER

Protein/peptide name IL-3
Protein synonyms MCGF|HCGF|Multi-CSF|P-cell stimulation factor|Interleukin-3
Environmentally Preferable
Protein/peptide type Recombinant
Arter Human
Kilde
Biologisk aktivitet The activity is determined by the dose-dependent stimulation of TF-1 cells which is typically observed at concentations <0,1 ng/ml.
Renhet >95% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
Formulering Lyophilized. Sterile filtered.
Molekylvekt ~15kDa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR