E-64

Leverandør: ENZO LIFE SCIENCES

ALX260007M005 ALX260007M025
ENZOALX260007M005EA 1425 NOK
ENZOALX260007M005 ENZOALX260007M025
E-64
Proteiner
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aff7a87f9663e6096b1ab9bdc7ee07198fce4e89&vid=28acfe09ea4d23e24365a6eb55f4f26eb9c87fdc&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Protein/peptide name E-64
Protein synonyms 3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
Environmentally Preferable
Arter
Kilde
CAS-nr
MDL No. MFCD00080261
Molekylformel C₁₅H₂₇N₅O₅
Lagringsforhold Freezer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR