Z-VAD-FMK
Leverandør: ENZO LIFE SCIENCES
ALX260020M001 ALX260020M005
ENZOALX260020M001EA 2450 NOK
ENZOALX260020M001 ENZOALX260020M005
Z-VAD-FMK
Proteiner
Caspase inhibitor
Irreversible, cell permeable broad-spectrum caspase inhibitor.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21782967a806888623cffa35edfc1bcb22e391a0&vid=6be0cd0c7ce932916047ee79ce47c51c959ad88d&prid=web&clid=SAPDAM