Ac-YVAD-CHO

Leverandør: ENZO LIFE SCIENCES

ALX260027M001 ALX260027M005
ENZOALX260027M001EA 1075 NOK
ENZOALX260027M001 ENZOALX260027M005
Ac-YVAD-CHO
Proteiner

Inhibitor of caspase-1 and -4


Potent and reversible inhibitor of caspase-1. Also inhibits caspase-4. Strongly inhibits anti-Fas induced apoptosis in L929-Fas cells.

Bestill nå

SPESIFIKASJONER

Protein/peptide name Ac-YVAD-CHO Ac-YVAD-CHO
Environmentally Preferable
Arter
Kilde
Renhet >96% (HPLC) >96% (HPLC)
Formulering Lyophilized __
Reconstitution instructions Add 0,2 ml DMSO per 1,0 mg peptide. Shake well until all the contents are dissolved. This furnishes a 10 mM solution of this material. __

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR