Ac-VDVAD-pNA

Leverandør: ENZO LIFE SCIENCES

ALX260059M001 ALX260059M005
ENZOALX260059M001EA 635 NOK
ENZOALX260059M001 ENZOALX260059M005
Ac-VDVAD-pNA
Proteiner

Caspase-2 substrate


Chromogenic substrate for caspase-2. Preferred substrate for caspase-2 (Km=53μM)

Bestill nå

SPESIFIKASJONER

Protein/peptide name Ac-VDVAD-pNA Ac-VDVAD-pNA
Environmentally Preferable
Arter
Kilde
Renhet >98% (HPLC) >98% (HPLC)
Formulering Lyophilized __

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR