Ac-VEID-CHO
Leverandør: ENZO LIFE SCIENCES
ALX260062M001 ALX260062M005
ENZOALX260062M001EA 1775 NOK
ENZOALX260062M001 ENZOALX260062M005
Ac-VEID-CHO
Proteiner
Inhibitor of caspase-6, -8 and -10
Reversible aldehyde inhibitor of caspase-6. Also inhibits caspase-8 and -10. Sequence is based on lamin A site of cleavage.