Ac-YVAD-AFC
Leverandør: ENZO LIFE SCIENCES
ALX260108M005 ALX260108M010
ENZOALX260108M005EA 2250 NOK
ENZOALX260108M005 ENZOALX260108M010
Ac-YVAD-AFC
Proteiner
Substrate for caspase-1 and -4
Fluorogenic substrate for caspase-1 and -4.