Z-WEHD-FMK
Leverandør: ENZO LIFE SCIENCES
ALX260140R020 ALX260140R100
ENZOALX260140R020EA 3225 NOK
ENZOALX260140R020 ENZOALX260140R100
Z-WEHD-FMK
Proteiner
Inhibitor of caspase-1, -4 and -5
Irreversible, cell permeable inhibitor of caspase-1 and -5. Inhibits also caspase-4. Designed as a methyl ester to facilitate cell permeability.