Z-VEID-FMK
Leverandør: ENZO LIFE SCIENCES
ALX260143R020 ALX260143R100
ENZOALX260143R020EA 3225 NOK
ENZOALX260143R020 ENZOALX260143R100
Z-VEID-FMK
Proteiner
Caspase-6 inhibitor
Irreversible, cell permeable inhibitor of caspase-6. The inhibitor is designed as a methyl ester to facilitate cell permeability.