Z-VEID-FMK

Leverandør: ENZO LIFE SCIENCES

ALX260143R020 ALX260143R100
ENZOALX260143R020EA 3225 NOK
ENZOALX260143R020 ENZOALX260143R100
Z-VEID-FMK
Proteiner

Caspase-6 inhibitor


Irreversible, cell permeable inhibitor of caspase-6. The inhibitor is designed as a methyl ester to facilitate cell permeability.

Bestill nå

SPESIFIKASJONER

Protein/peptide name Z-VEID-FMK Z-VEID-FMK
Environmentally Preferable
Arter
Kilde
Renhet >98% (HPLC) >98% (HPLC)
Formulering Liquid. In DMSO. __

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR