AM 281 98 HPLC_ASSAY_METHOD

Leverandør: ENZO LIFE SCIENCES
Danger

ALX270240M005
ENZOALX270240M005EA 1475 NOK
ENZOALX270240M005
AM 281 98 HPLC_ASSAY_METHOD
AM 281

CB1 receptor ligand.


Analog of the cannabinoid receptor (CB) antagonist SR 141716A. Potent CB1 receptor antagonist/inverse agonist (Ki = 14 nM).


Soluble in methanol (1 mg/ml) or DMSO (2 mg/ml at RT, 50 mg/ml in hot DMSO, after cooling this solution is stable over several days).

Formel: C₂₁H₁₉Cl₂IN₄O₂
MW: 557.2 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 202463-68-1
FN: 2811
ADR: 6.1 III

Bestill nå

Spesifikasjon Testresultater

Purity ≥98% (HPLC)
Identity Determined by ¹H-NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR