Pifithrin-α (cyclic) hydrobromide 95 HPLC_ASSAY_METHOD
Leverandør: ENZO LIFE SCIENCES
ALX270287M010 ALX270287M050
ENZOALX270287M010EA 2850 NOK
ENZOALX270287M010 ENZOALX270287M050
Pifithrin-α (cyclic) hydrobromide 95 HPLC_ASSAY_METHOD
Pifithrin-α (cyclic) hydrobromide
p53 inhibitor
Stable analog of pifithrin-alpha (Prod. No. BML-GR325), with biological properties similar to its parent compound, but with reduced cytotoxicity. Inhibitor of p53. Reversibly inhibits p53-dependent transactivation of p53-responsive genes and reversibly blocks p53-mediated apoptosis. Potent STAT6 transcriptional inhibitor.
CAS-nummer: 60477-34-1