D-erythro-Sphingosine (synthetic) ≥99%

Leverandør: ENZO LIFE SCIENCES

Synonymer: D-Sphingosine, trans-D-erythro-2-Amino-4-octadecene-1,3-diol, (2S,3R,E)-2-Amino-4-octadecene-1,3-diol, 4-Sphingenine, (2S,3R,E)-2-Aminooctadec-4-ene-1,3-diol

ALX306011M025
ENZOALX306011M025EA 1225 NOK
ENZOALX306011M025
D-erythro-Sphingosine (synthetic) ≥99%
D-erythro-Sphingosine

PKC inhibitor


Common component of cell membranes, often as phosphatidyl conjugates. Inhibitor of protein kinase C (PKC) and calmodulin-dependent enzymes.

Formel: C₁₈H₃₇NO₂
MW: 299.50 g/mol
MDL nummer: MFCD00036751
CAS-nummer: 123-78-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR