D-erythro-Sphingosine-1-phosphate (caged) ≥95%

Leverandør: ENZO LIFE SCIENCES
Warning

Synonymer: 1-SPP (caged)

ALX306017C500
ENZOALX306017C500EA 3575 NOK
ENZOALX306017C500
D-erythro-Sphingosine-1-phosphate (caged) ≥95%
D-erythro-Sphingosine-1-phosphate (caged)

A photolysable derivative of sphingosine 1-phosphate (1-SPP). Important tool for the study of 1-SPP mediated intracellular events.


Hygroscopic and soluble in methanol or 100% ethanol.

Bestill nå

Spesifikasjon Testresultater

Purity ≥95%
Source Synthetic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR