Okadaic acid

Leverandør: ENZO LIFE SCIENCES

ALX350003C025 ALX350003C050 ALX350003C100 ALX350003M001
ENZOALX350003C025EA 1650 NOK
ENZOALX350003C025 ENZOALX350003C050 ENZOALX350003C100 ENZOALX350003M001
Okadaic acid
Okadaic acid
Formel: C₄₄H₆₈O₁₃
MDL nummer: MFCD00083455
CAS-nummer: 78111-17-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR