Dihydrocapsaicin

Leverandør: ENZO LIFE SCIENCES

Synonymer: N-(4-Hydroxy-3-methoxybenzyl)-8-methylnonanamide, 8-Methyl-N-vanillylnonanamid

ALX350052M050
ENZOALX350052M050EA 1650 NOK
ENZOALX350052M050
Dihydrocapsaicin
Dihydrocapsaicin
Formel: C₁₈H₂₉NO₃
MDL nummer: MFCD00057794
CAS-nummer: 19408-84-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR