Topotecan hydrochloride 98 HPLC_ASSAY_METHOD

Leverandør: ENZO LIFE SCIENCES

Synonymer: SKF-104864A hydrochloride, 10-[(Dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride, NSC 609669 hydrochloride, Topotecan HCl, 9-[(dimethylamino)methyl]-10-hydroxy-(20S)-camptothecin hydrochloride, (S)-10-((Dimethylamino)methyl)-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride

ALX350133M001 ALX350133M005 ALX350133M025
ENZOALX350133M001EA 620 NOK
ENZOALX350133M001 ENZOALX350133M005 ENZOALX350133M025
Topotecan hydrochloride 98 HPLC_ASSAY_METHOD
Topotecan hydrochloride

Potent antitumor agent. Inhibitor of DNA-topoisomerase. Analog of (S)-(⁺)-camptothecin. Used in chemotherapy of several different cancers.


  • Topoisomerase I inhibitor


Light yellow to greenish powder.


Soluble in water (1mg/ml) or methanol.

Formel: C₂₃H₂₃N₃O₅.HCl
MW: 457.91 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00866235
CAS-nummer: 119413-54-6
New Product

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR