Daunorubicin hydrochloride (from Streptomyces species)

Leverandør: ENZO LIFE SCIENCES

ALX380043M010
ENZOALX380043M010EA 2350 NOK
ENZOALX380043M010
Daunorubicin hydrochloride (from Streptomyces species)
Daunorubicin hydrochloride

Antitumor antibiotic.


Anthracycline anticancer antibiotic. Induces DNA damage by intercalating into DNA and inhibiting topoisomerase I and II. Inhibits RNA and DNA synthesis. Induces DNA single strand breaks and apoptosis in tumor cells.


Orange to red crystalline powder


Soluble in water or methanol


Forpakning: Shipping temperature: Ambient

Formel: C₂₇H₂₉NO₁₁·HCl
Lagringstemperatur: Fryseskap
MDL nummer: MFCD04974507
CAS-nummer: 23541-50-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR