Cytochalasin A

Leverandør: ENZO LIFE SCIENCES

ALX380057M001 ALX380057M005 ALX380057M010
ENZOALX380057M001EA 1200 NOK
ENZOALX380057M001 ENZOALX380057M005 ENZOALX380057M010
Cytochalasin A
Cytochalasin A
Formel: C₂₉H₃₅NO₅
MW: 477.6 g/mol
Smeltepunkt: 182…185 °C
MDL nummer: MFCD00005935
CAS-nummer: 14110-64-6
EINECS: 237-964-9
FN: 1544
ADR: 6.1 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR