Blasticidin S hydrochloride (from Streptomyces griseochromogenes) ≥98% (by HPLC)

Leverandør: ENZO LIFE SCIENCES

ALX380089M100
ENZOALX380089M100EA 6025 NOK
ENZOALX380089M100
Blasticidin S hydrochloride (from Streptomyces griseochromogenes) ≥98% (by HPLC)
Blasticidin S hydrochloride

Antibiotic.


Inhibitor of DNA and protein synthesis.


White to off-white powder


Soluble in water, insoluble in organic solvents


Forpakning: Shipping temperature: Ambient

Formel: C₁₇H₂₇ClN₈O₅
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD02091640
CAS-nummer: 3513-03-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR