Carbenicillin disodium salt ≥92%

Supplier: ENZO LIFE SCIENCES
Danger

ALX380270G001 ALX380270M250
ENZOALX380270G001EA 2000 NOK
ENZOALX380270G001 ENZOALX380270M250
Carbenicillin disodium salt ≥92%
Carbenicillin disodium salt

Inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Effective against Gram-positive and Gram-negative bacteria and Pseudomonas.

Formula: C₁₇H₁₆N₂O₆S·2Na
Storage Temperature: Fryseskap
MDL Number: MFCD00077683
CAS Number: 4800-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR