Carboplatin ≥98% (by HPLC)

Leverandør: ENZO LIFE SCIENCES
Danger

Synonymer: cis-Diammine(1,1-cyclobutanedicarboxylato)platinum(II), cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II), 1,1-Cyclobutanedicarboxylatodiammineplatinum(II)

ALX400041M025 ALX400041M100
ENZOALX400041M025EA 800 NOK
ENZOALX400041M025 ENZOALX400041M100
Carboplatin ≥98% (by HPLC)
Carboplatin

Antitumor agent.


An analog of cisplatin.


Soluble in water.

Formel: C₆H₁₂N₂O₄Pt
MW: 371.26 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00070464
CAS-nummer: 41575-94-4
EINECS: 255-446-0
Merck Indeks: 14,01822

Bestill nå

Spesifikasjon Testresultater

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR