Spermine NONOate ≥97% (by NMR)

Leverandør: ENZO LIFE SCIENCES
Warning

ALX4300135005 ALX430013M005 ALX430013M025
ENZOALX4300135005EA 2525 NOK
ENZOALX4300135005 ENZOALX430013M005 ENZOALX430013M025
Spermine NONOate ≥97% (by NMR)
Spermine NONOate

Nitric oxide (NO) donor.


A convenient reagent for preparing aqueous solutions of NO. Relatively stable in alkaline solution.


Hygroscopic. Soluble in water, methanol or 100% ethanol.


NB: Protect from light and oxygen. Keep under inert gas.

Formel: C₁₀H₂₆N₆O₂
Lagringstemperatur: Ultra lav temperatur fryser (–80°C)
CAS-nummer: 136587-13-8

Bestill nå

Spesifikasjon Testresultater

Purity ≥97% (NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR