DETA NONOate (NOC-18) 97 1H-NMR

Leverandør: ENZO LIFE SCIENCES
Warning

Synonymer: DETA/NO, 2,2′-(Hydroxynitrosohydrazino)bis-ethanamine, Diethylenetriamine NONOate, 3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene

ALX4300145005 ALX430014M005 ALX430014M025
ENZOALX4300145005EA 2450 NOK
ENZOALX4300145005 ENZOALX430014M005 ENZOALX430014M025
DETA NONOate (NOC-18) 97 1H-NMR
DETA NONOate (NOC-18)

Nitric oxide (NO) donor.


Induces apoptosis in macrophages. The half-life time of DETA NONOate is 20 hours at 37 °C and 56 hours at 22 - 25 °C in 0.1 M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0. DETA NONOate liberates 2 mol of NO per mol of parent compound.


Soluble in water or ethanol. Relatively stable in alkaline solution.


NB: Protect from light and oxygen.

Formel: C₄H₁₃N₅O₂
MW: 163.181 g/mol
Lagringstemperatur: Ultra lav temperatur fryser (–80°C)
MDL nummer: MFCD00278808
CAS-nummer: 146724-94-9

Bestill nå

Spesifikasjon Testresultater

Purity ≥97% (1H-NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR