MAHMA NONOate 98

Leverandør: ENZO LIFE SCIENCES
Warning

ALX4300155005 ALX430015M005 ALX430015M025
ENZOALX4300155005EA 2675 NOK
ENZOALX4300155005 ENZOALX430015M005 ENZOALX430015M025
MAHMA NONOate 98
MAHMA NONOate

Nitric oxide (NO) donor.


The half-life time of MAHMA NONOate is 2.7 min. at 22 °C in 0.1 M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0.


Soluble in water. Relatively stable in alkaline solution.


NB: Protect from light and moisture.

Formel: C₈H₂₀N₄O₂
MW: 204.3 g/mol
Lagringstemperatur: Fryseskap
CAS-nummer: 146724-86-9

Bestill nå

Spesifikasjon Testresultater

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR