PAPA NONOate 97

Leverandør: ENZO LIFE SCIENCES
Warning

ALX4300165005 ALX430016M005 ALX430016M025
ENZOALX4300165005EA 2675 NOK
ENZOALX4300165005 ENZOALX430016M005 ENZOALX430016M025
PAPA NONOate 97
PAPA NONOate

Nitric oxide (NO) donor.


The half-life time of PAPA NONOate is 76.6 min at 22 °C in 0.1 M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0.


Soluble in water or methanol. Relatively stable in alkaline solution.


NB: Protect from moisture. Keep under inert gas.

Formel: C₆H₁₆N₄O₂
MW: 176.2 g/mol
Lagringstemperatur: Fryseskap
CAS-nummer: 146672-58-4

Bestill nå

Spesifikasjon Testresultater

Purity ≥97%
Identity Determined by IR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR