DEA NONOate 98 NMR_ASSAY_METHOD, solid_white to off white

Leverandør: ENZO LIFE SCIENCES
Danger

Synonymer: 2-(N,N-diethylamino)diazenolate-2-oxide diethylammonium salt

Inert_Gas_filled

ALX4300345005 ALX430034M010 ALX430034M050
ENZOALX4300345005EA 1750 NOK
ENZOALX4300345005 ENZOALX430034M010 ENZOALX430034M050
DEA NONOate 98 NMR_ASSAY_METHOD, solid_white to off white
DEA NONOate

DEA NONOate is used as a nitric oxide donor.


Hygroscopic and soluble in 100% ethanol (25 mg/ml), DMSO (2 mg/ml), dimethyl formamide (2 mg/ml) or PBS, pH 7.2 (10 mg/ml).

Formel: C₄H₁₀N₃O₂·C₄H₁₂N
MW: 206.2 g/mol
Lagringstemperatur: Ultra lav temperatur fryser (–80°C)
CAS-nummer: 372965-00-9

Bestill nå

Spesifikasjon Testresultater

Purity (NMR) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR