DPTA NONOate 98 NMR_ASSAY_METHOD

Leverandør: ENZO LIFE SCIENCES
Warning

Synonymer: Dipropylenetriamine NONOate

ALX4300665005 ALX430066M005 ALX430066M025
ENZOALX4300665005EA 3075 NOK
ENZOALX4300665005 ENZOALX430066M005 ENZOALX430066M025
DPTA NONOate 98 NMR_ASSAY_METHOD
DPTA NONOate

Nitric oxide (NO) donor.


The half-life time is 5 hours at 22 °C in 0.1 M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0.


Highly soluble in water.

Formel: C₆H₁₇N₅O₂
MW: 191.2 g/mol
Lagringstemperatur: Ultra lav temperatur fryser (–80°C)
CAS-nummer: 146724-95-0

Bestill nå

Spesifikasjon Testresultater

Purity (NMR) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR