DL-α-Tocopherol (Vitamin E)

Leverandør: ENZO LIFE SCIENCES

Synonymer: (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E

ALX460018G001
ENZOALX460018G001EA 595 NOK
ENZOALX460018G001
DL-α-Tocopherol (Vitamin E)
DL-α-Tocopherol (Vitamin E)
Formel: C₂₉H₅₀O₂
MW: 430.71 g/mol
MDL nummer: MFCD00072051
CAS-nummer: 10191-41-0
EINECS: 233-466-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR