Adenosine-3',5'-cyclic monophosphate sodium salt ≥98% (by HPLC)

Leverandør: ENZO LIFE SCIENCES

Synonymer: 3′,5′-Cyclic AMP sodium salt, 3',5'-Cyclic AMP Sodium Salt, Adenosine 3',5'-Cyclic Monophosphate Sodium Salt Hydrate, 3',5'-cAMP Na, (4aR,6R,7R,7aS)-6-(6-Amino-9H-purin-9-yl)-2,7-dihydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinine 2-oxide, sodium salt, cAMP-Na, 3',5'-Cyclic AMP Sodium Salt Hydrate, 3',5'-cAMP Na Hydrate

ALX480011M100
ENZOALX480011M100EA 970 NOK
ENZOALX480011M100
Adenosine-3',5'-cyclic monophosphate sodium salt ≥98% (by HPLC)
Adenosine-3',5'-cyclic monophosphate sodium salt

PKA activator.


Key regulator of many cellular reactions. Naturally occurring activator of cAMP-dependent protein kinase (PKA).


Soluble in water.

Formel: C₁₀H₁₁N₅NaO₆P
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00069736
CAS-nummer: 37839-81-9
Merck Indeks: 14,02708
New Product

Bestill nå

Spesifikasjon Testresultater

Purity (HPLC) ≥98%
Identity Identity determined by ³¹P- NMR, ¹H-NMR, FAB-MS and UV

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR