8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate triethylammonium salt ≥99% (by HPLC)

Leverandør: ENZO LIFE SCIENCES

ALX480031M005
ENZOALX480031M005EA 3300 NOK
ENZOALX480031M005
8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate triethylammonium salt ≥99% (by HPLC)
8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate triethylammonium salt

Activator of PKG and CNG channels.


Selective activator of cGMP-dependent protein kinase Iα, Iβ and type II as well as of cGMP-gated ion channels.


Higher activation potential as compared to cGMP; excellent cell membrane permeability and phosphodiesterase stability. Preferred alternative to unsuitable analogs such as dibutyryl-cGMP.


Soluble in water.

Formel: C₁₆H₁₄ClN₅O₇PS·C₆H₁₆N
MW: 589 g/mol
Lagringstemperatur: Fryseskap
CAS-nummer: 54364-02-2

Bestill nå

Spesifikasjon Testresultater

Purity (HPLC) ≥99%
Identity Determined by MS and UV


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR