Human recombinant TRAIL (soluble) (from E. coli)
Leverandør: ENZO LIFE SCIENCES
ALX5220033010 ALX522003C010
ENZOALX5220033010EA 8150 NOK
ENZOALX5220033010 ENZOALX522003C010
Human recombinant TRAIL (soluble) (from E. coli)
Proteiner
Produced in E. coli. The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.