Human recombinant TRAILR2:Fc, FITC (Fluorescein Isothiocyanate)
Leverandør: ENZO LIFE SCIENCES
ALX522005FC050
ENZOALX522005FC050EA 8150 NOK
ENZOALX522005FC050
Human recombinant TRAILR2:Fc, FITC (Fluorescein Isothiocyanate)
Proteiner
Produced in HEK 293 cells. The extracellular domain of human TRAIL-R2 (DR5) (aa 52-212) is fused to the Fc portion of human IgG1.