Human recombinant Fc:CD40L (soluble)
Leverandør: ENZO LIFE SCIENCES
ALX522076C010
ENZOALX522076C010EA 7450 NOK
ENZOALX522076C010
Human recombinant Fc:CD40L (soluble)
Proteiner
Produced in HEK 293 cells. The extracellular domain of human CD40L (CD154) (aa 116-261) is fused at the N-terminus to the Fc portion of human IgG1 and a linker peptide (20 aa).