Human recombinant TL1A (soluble) (from E. coli)
Leverandør: ENZO LIFE SCIENCES
ALX522084C010
ENZOALX522084C010EA 7325 NOK
ENZOALX522084C010
Human recombinant TL1A (soluble) (from E. coli)
Proteiner
Produced in E. coli. The extracellular domain of human TL1A (aa 93-251) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.
New Product
Bestill nå
SPESIFIKASJONER
Protein/peptide name TL1A (soluble)
Protein/peptide type Recombinant
Arter Human
Kilde E. coli
Renhet >85% (SDS-PAGE)
Konsentrasjon 0,1 mg/ml after reconstitution
Formulering Lyophilised. Contains PBS.
Reconstitution instructions Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.