SNC80 ≥98% (by HPLC), white to off-white solid

Leverandør: ENZO LIFE SCIENCES

ALX550172M005 ALX550172M025
ENZOALX550172M005EA 1475 NOK
ENZOALX550172M005 ENZOALX550172M025
SNC80 ≥98% (by HPLC), white to off-white solid
SNC80

delta-opioid receptor agonist


Highly selective and potent non-peptide δ-opioid receptor agonist.


Soluble in 100% ethanol, methanol, DMSO or dichloromethane.

Formel: C₂₈H₃₉N₃O₂
MW: 449.6 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 156727-74-1

Bestill nå

Spesifikasjon Testresultater

Purity (HPLC) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR