5-Nitro-6,7-dichloro-1,4-dihydro-2,3-quinoxalinedione ≥98% (by TLC)

Leverandør: ENZO LIFE SCIENCES

ALX550228M005 ALX550228M025
ENZOALX550228M005EA 2025 NOK
ENZOALX550228M005 ENZOALX550228M025
5-Nitro-6,7-dichloro-1,4-dihydro-2,3-quinoxalinedione ≥98% (by TLC)
5-Nitro-6,7-dichloro-1,4-dihydro-2,3-quinoxalinedione

Potent and selective NMDA receptor glycine site antagonist.


Soluble in DMSO or dilute aqueous base.

Formel: C₈H₃Cl₂N₃O₄
MW: 276.0 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 153504-81-5

Bestill nå

Spesifikasjon Testresultater

Purity (TLC, elemental analysis) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR