Trifluoperazine dihydrochloride ≥98%

Leverandør: ENZO LIFE SCIENCES

Synonymer: 10-[3-(4-Methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine Dihydrochloride

ALX550310G005
ENZOALX550310G005EA 2000 NOK
ENZOALX550310G005
Trifluoperazine dihydrochloride ≥98%
Trifluoperazine dihydrochloride

Antipsychotic


Facilitates agonist induced elevation of cytosolic calcium at low concentrations (10µM). Antagonizes calmodulin at higher concentrations. Antipsychotic and sedative dopamine antagonist. Potent and irreversible inhibitor of cAMP-gated cationic channels. Inhibits DNA repair. Cell permeable. Potent neurotoxin.

Formel: C₂₁H₂₄F₃N₃S
MW: 480.42 g/mol
MDL nummer: MFCD00012656
CAS-nummer: 440-17-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR