Lipopolysaccharides (from E.coli) serotype O55:B5, TLRgrade™ ready-to-use

Leverandør: ENZO LIFE SCIENCES
Danger

ALX581013L001 ALX581013L002
ENZOALX581013L001EA 2625 NOK
ENZOALX581013L001 ENZOALX581013L002
Lipopolysaccharides (from E.coli) serotype O55:B5, TLRgrade™ ready-to-use
Lipopolysaccharides

TLR4 activator.


Smooth (S)-form LPS isolated and purified from E. coli 055:B5 by a modification of the phenol water extraction and PCP extraction method, converted to the uniform sodium salt form.


Specific activator of Toll-like receptor (TLR) 4. Does not activate TLR2 or other TLRs as determined with splenocytes and macrophages from TLR4 deficient mice. Smooth (S)-form LPS are commonly the preferred choice for whole animal studies, whereas Rough (R)-form LPS are primarily used in cellular in vitro activation studies.

Bestill nå

Spesifikasjon Testresultater

Concentration 1 mg/ml
Formulation Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
Purity Absence of detectable protein or DNA contaminants with agonistic TLR activity

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR