Lipid A 1 mg/ml in water (from E.coli), Serotype R515 (Re) ≥99.9% serotype J5 (Rc), TLRgrade™ ready-to-use non pyrogenic, sterile

Leverandør: ENZO LIFE SCIENCES
Danger

ALX581200L001 ALX581200L002
ENZOALX581200L001EA 4400 NOK
ENZOALX581200L001 ENZOALX581200L002
Lipid A 1 mg/ml in water (from E.coli), Serotype R515 (Re) ≥99.9% serotype J5 (Rc), TLRgrade™ ready-to-use non pyrogenic, sterile
Lipid A

TLR4 activator.


Prepared by mild acid hydrolysis from purified E. coli R515 (Re) LPS (isolated by a modification of the PCP extraction method), converted to the uniform salt form.


Activates Toll-like receptor (TLR) 4. Does not activate TLR2 or other TLRs as determined with splenocytes and macrophages from TLR4 deficient mice.


Bestillingsinformasjon: Shipped at ambient temperature

Bestill nå

Spesifikasjon Testresultater

Concentration 1 mg/ml
Formulation Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
Purity ≥99.9% (regarding protein or DNA contaminants, KDO not detectable)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR