DAR-4M AM (cell permeable) 5 mM 98 HPLC_ASSAY_METHOD

Leverandør: ENZO LIFE SCIENCES
Warning

Synonymer: Diaminorhodamine-4M AM

ALX620069M001
ENZOALX620069M001EA 11550 NOK
ENZOALX620069M001
DAR-4M AM (cell permeable) 5 mM 98 HPLC_ASSAY_METHOD
DAR-4M AM (cell permeable)

Fluorescent probe for NO detection. Cell permeable.


Cell permeable analog of DAR-4M. Can be used to detect nitric oxide (NO) directly in cell specimens prepared from various sources, such as the brain (Ex: ~560 nm; Em: 575 nm). Stable in a pH range of 4 – 12. Low decrease in fluorescence intensity over time. Detection limit 7 nM.

Bestill nå

Spesifikasjon Testresultater

Concentration ~5 mM
Purity ≥98% (HPLC)
Formulation Dissolved in 0.32 ml DMSO


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR